Hydrobromic acid, Alfa Aesar™

Manufacturer: Alfa Aesar 14036AZ

Quantity 4 x 940mL

 View more versions of this product

Catalog No. AA14036AZ

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10035-10-6
    Physical Form Liquid
    Molecular Formula HBr
    Formula Weight 80.92
    MDL Number MFCD00011323
    Synonym Hydrogen bromide
    Chemical Name or Material Hydrobromic acid

    Specifications

    Formula Weight 80.92
    Quantity 4 x 940mL
    EINECS Number 233-113-0
    Physical Form Liquid
    Density 1.49g/mL
    Molecular Formula HBr
    MDL Number MFCD00011323
    CAS 10035-10-6
    Synonym Hydrogen bromide
    Chemical Name or Material Hydrobromic acid

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit