Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
10035-10-6,7732-18-5
BrH
80.912
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide
260
CHEBI:47266
bromane
Br
10035-10-6,7732-18-5
BrH
80.912
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide
260
CHEBI:47266
bromane
Br
C-X bond formation reagent
Color Brown to Yellow
Quantity 1L
Linear Formula HBr
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Merck Index 15, 4816
Density 1.4800g/mL
Formula Weight 80.9
Physical Form Solution
Percent Purity ≥47%
Grade Pure
Packaging Glass bottle
Solubility Solubility in water: soluble.
Specific Gravity 1.48
Storage Information Slowly darkens on exposure to air and light
Melting Point -11.0°C
Boiling Point 126.0°C to 128.0°C
Chemical Name or Material Hydrobromic acid, tech.
Show More Show Less

GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

EINECSNumber : 233-113-0

RTECSNumber : MW3850000

TSCA : TSCA