Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrobromic Acid, purum p.a., Honeywell™
Click to view available options
Quantity:
250 mL
1 L
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Density | 1.49g/mL at 25°C |
Vapor Pressure | 8mmHg (25°C), 320psi (21.1°C) |
Quantity | 250 mL |
UN Number | 1788 |
Beilstein | 3587158 |
Formula Weight | 80.91g/mol |
Percent Purity | ≥48% |
Grade | purum p.a. |
Chemical Name or Material | Hydrobromic Acid |
Safety and Handling
Hazard Statement | P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
Precautionary Statement | H314-H335 |
EINECSNumber | 233-113-0 |
RTECSNumber | MW3850000 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction