Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Hydrobromic Acid Solution, purum p.a., ≥62% (T), Honeywell™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass Bottle
10035-10-6
BrH
80.912
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide
260
CHEBI:47266
bromane
Br
10035-10-6
BrH
80.912
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide
260
CHEBI:47266
bromane
Br

Purum p.a.,≥62% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Color Brown-Yellow
Quantity 1L
UN Number UN1788
Beilstein 3587158
Density 1.73g/cu.cm
Formula Weight 80.91g/mol
Percent Purity ≥62% (T)
Grade purum p.a.
Packaging Glass Bottle
Physical State Clear liquid
Melting Point -11°C
Boiling Point 126°C
Chemical Name or Material Hydrobromic Acid Solution
Show More Show Less

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H314-H335

DOTInformation : 8/PGII