Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid 0.01N DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

Click to view available options
Quantity:
230.8mL
1.9L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle
7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
Quantity 1.9L
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Hydrochloric Acid 0.01N DILUT-IT™ Dissolution Media Concentrate