Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.01N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
chlorane
Cl

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Formula Weight 36.46
Packaging Cubetainer
Specific Gravity 1L = 1.00kg
Chemical Name or Material Hydrochloric Acid, 0.01N Volumetric Solution
Grade BAKER ANALYZED™ Reagent