Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, 0.1 N standard solution, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1L
2.5L
10L
25L
Packaging:
Glass bottle
Plastic Drum
Plastic bottle
7647-01-0,7732-18-5
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0,7732-18-5
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
Volumetric standard solution

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Color Colorless
Quantity 1L
Additional Information Vapor Pressure: 14 mmHg at 20°C
Linear Formula HCl
Fieser 04,450; 05,533; 06,283
Merck Index 15, 4818
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene
Concentration 0.0998 to 0.1002N (20°C)
Density 1.0000g/mL
Formula Weight 36.46
Physical Form Liquid
Packaging Glass bottle
Specific Gravity 1
Boiling Point 100.0°C
Chemical Name or Material Hydrochloric acid, 0.1 N Standard Solution
Show More Show Less

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health: 2
Flammability: 0
Instability: 0

SIGNAL WORD: CAUTION!