Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, 0.1 N standard solution, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
25L
Packaging:
Glass bottle
Plastic Drum
Plastic bottle
7647-01-0,7732-18-5
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0,7732-18-5
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
Volumetric standard solution
Color Colorless
Quantity 2.5L
Additional Information Vapor Pressure: 14 mmHg at 20°C
Linear Formula HCl
Fieser 04,450; 05,533; 06,283
Merck Index 15, 4818
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene
Concentration 0.0998 to 0.1002N (20°C)
Density 1.0000g/mL
Formula Weight 36.46
Physical Form Liquid
Packaging Plastic bottle
Solubility Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene
Specific Gravity 1
Boiling Point 100.0°C
Chemical Name or Material Hydrochloric acid, 0.1 N Standard Solution
Show More Show Less

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health: 2
Flammability: 0
Instability: 0

SIGNAL WORD: CAUTION!