Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.1 Normal (N/10) in Isopropanol with Bromophenol Blue, Reagents

Catalog No. 01335083
Click to view available options
Quantity:
100 mL
1 L

Hydrochloric Acid, 0.1 Normal (N/10) in Isopropanol with Bromophenol Blue, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Hydrochloric Acid, 0.1 Normal (N/10) in Isopropanol with Bromophenol Blue
pH 1
Color Yellow
CAS 67-63-0, 7732-18-5
Boiling Point 83°C
Packaging Glass Bottle
Specific Gravity 0.8
Quantity 1 L
Molecular Formula HCl/C3H7OH
Physical Form Liquid
Synonym Hydrogen chloride solution
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.