Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric Acid, 0.100 N (N/10), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Specifications
Specifications
| CAS Min % | 97.0, 0.95 |
| CAS Max % | 100.0, 0.99 |
| Packaging | Natural Poly Bottle |
| Quantity | 1 L |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 0.100N |
| Chemical Name or Material | Hydrochloric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction