Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 1.0 N Aqueous Solution, APHA, 1 N +/-0.005 N, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
7647-01-0,7732-18-5
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
[H+].[Cl-]
7647-01-0,7732-18-5
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
[H+].[Cl-]

H-135, 7647-01-0

Spectrum™ Hydrochloric Acid, 1.0 N Aqueous Solution, APHA , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.
Quantity 1 L
CAS Max % 0.036,0.964
UN Number UN1789
Concentration 1 N +/-0.005
Packaging Poly Bottle