Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 1.000 +/- 0.001 N, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner R370000012X500

Packaging Poly natural
Quantity 6L

 View more versions of this product

Catalog No. 37-001-6CS


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

7647-01-0,7732-18-5
1.000 ± 0.001 N at 20°C
Hydrochloric Acid, 1 N

Specifications

6L
1.000 ± 0.001 N at 20°C
Poly natural
Hydrochloric Acid, 1 N
7647-01-0,7732-18-5
Odorless to slightly pungent
Aqueous Solution
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit