Hydrochloric Acid, 1.000 +/- 0.001 N, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner R37000004X4A

Packaging Poly natural
Quantity 16L

 View more versions of this product

Catalog No. 37-001-CS

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7647-01-0,7732-18-5
    Assay Percent Range 1.000 ± 0.001 N at 20°C
    Chemical Name or Material Hydrochloric Acid, 1 N

    Specifications

    Quantity 16L
    Solvent or Matrix Aqueous Solution
    Assay Percent Range 1.000 ± 0.001 N at 20°C
    CAS 7647-01-0,7732-18-5
    Odor Odorless to slightly pungent
    Packaging Poly natural
    Chemical Name or Material Hydrochloric Acid, 1 N

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit