Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 1% (v/v) Aqueous Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
7647-01-0,7732-18-5
7647-01-0,7732-18-5
Quantity 4 L
Grade Chemical Solution
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.