Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 10% (v/v; 1+9) Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
7647-01-0
7647-01-0
Quantity 4 L
Grade Chemical Solution
Packaging Poly Bottle