Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 20° Be, Technical, Approx. 30%, Spectrum™

Click to view available options
Quantity:
4 L
20 L
4 x 4 L
55 gal.
Packaging:
Drum
Poly Drum
Poly-Coated Glass Bottle
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
[H+].[Cl-]
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
[H+].[Cl-]

H1040, 7647-01-0, HCl

Spectrum™ Hydrochloric Acid, 20 DEG Be, Technical is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.
Quantity 55 gal.
CAS Max % 1
UN Number UN1789
Percent Purity Approx. 30%
Packaging Drum