Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 36.5 to 38.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
Glass Bottle
Poly Bottle
PVC Coated Glass Bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
chlorane
Cl

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
Formula Weight 36.46
Packaging Glass Bottle
Specific Gravity 1L = 1.18kg
Chemical Name or Material Hydrochloric Acid
Assay 36.5 to 38.0%
Grade Baker Analyzed™ ACS Reagent