Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 36.5 to 38.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
Glass Bottle
Poly Bottle
PVC Coated Glass Bottle
7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
Formula Weight 36.46
Percent Purity 36.5 to 38.0%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.18kg
Chemical Name or Material Hydrochloric Acid