Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5.0 Normal Standardized, NIST Traceable, Reagents

Catalog No. 01335124
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L

Catalog No. 01-335-124

Supplier: Reagents Holdings Llc CH101400500A

Only null left
Add to Cart
Add to Cart

Hydrochloric Acid, 5.0 Normal Standardized, NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Green/Yellow
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0843
Quantity 500 mL
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 5.0 Normal Standardized, NIST Traceable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.