Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5.0 Normal Standardized, NIST Traceable, Reagents

Catalog No. 01335126
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Catalog No. Quantity
01-335-126 20 L
01-335-128 1 L
01-335-127 2.5 L
01-335-125 4 L
01-335-124 500 mL
5 options

Catalog No. 01-335-126

Supplier: Reagents Holdings Llc CH10140020F

Only null left
Add to Cart
Add to Cart

Hydrochloric Acid, 5.0 Normal Standardized, NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Green/Yellow
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0843
Quantity 20 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 5.0 Normal Standardized, NIST Traceable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.