Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 6.0N Solution, Biotech Reagent, J.T.Baker™

Click to view available options
Quantity:
19L
1L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Quantity 19L
CAS 7647-01-0
Molecular Formula ClH
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.458
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46
Packaging Poly Bottle
Specific Gravity 1L = 1.10kg
Chemical Name or Material Hydrochloric Acid, 6.0N Solution
Grade Biotech Reagent Grade
Show More Show Less