Hydrochloric acid, 6.0N Standardized Solution, Alfa Aesar™

Manufacturer: Alfa Aesar 44921K7

Quantity 4L

 View more versions of this product

Catalog No. AA44921K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-01-0,7732-18-5
    Physical Form Liquid
    Molecular Formula HCl
    Formula Weight 36.46
    MDL Number MFCD00011324
    Chemical Name or Material Hydrochloric acid

    Specifications

    Physical Form Liquid
    Formula Weight 36.46
    Quantity 4L
    Color Colorless to pale yellow
    DOT Information Hazard Class: 8, Packing Group: II
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    EINECS Number 231-595-7
    Appearance Colorless to pale yellow liquid
    Molecular Formula HCl
    MDL Number MFCD00011324
    CAS 7647-01-0,7732-18-5
    Concentration or Composition (by Analyte or Components) Hydrochloric acid: 49.6%; Water: 50.4%
    Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Hydrochloric acid

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit