Hydrochloric acid, 99.999% (metals basis), 36.5% min., Alfa Aesar™

Manufacturer: Alfa Aesar 87617K4

Quantity 2.5L

 View more versions of this product

Catalog No. AA87617K4

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7647-01-0
    Physical Form Clear colorless liquid
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula HCl
    Formula Weight 36.46g/mol
    MDL Number MFCD00011324
    Chemical Name or Material Hydrochloric acid

    Specifications

    Appearance Colorless liquid
    Odor Pungent
    Color Colorless
    Quantity 2.5L
    CAS 7647-01-0
    Physical Form Clear colorless liquid
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula HCl
    Formula Weight 36.46g/mol
    MDL Number MFCD00011324
    Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
    DOT Information Hazard Class: 8, Packing Group: II
    EINECS Number 231-595-7
    Chemical Name or Material Hydrochloric acid

    H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit