Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrochloric acid, Acculute Std. Volumetric Soln., Final Concentration 1.0N, Thermo Scientific™
Click to view available options
Quantity:
1 Unit
6 Units
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 1 Unit |
Color | Colorless |
IUPAC Name | palladium(2+) dichloride |
Formula Weight | 36.46 |
Odor | Acidic |
Physical Form | Liquid |
Chemical Name or Material | Hydrochloric acid |
Safety and Handling
Hazard Category | H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation. |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
DOTInformation | Hazard Class: 8, Packing Group: II |
EINECSNumber | 231-595-7 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction