Hydrochloric acid, AR Select™ Plus, Macron Fine Chemicals™

macron

Manufacturer: Macron V07846

Packaging SAFEMOR Container
Quantity 4L

 View more versions of this product

Catalog No. 14-650-512

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7647-01-0
    Physical Form Liquid
    Molecular Formula HCl
    Formula Weight 36.46
    Synonym Muriatic acid solution
    Chemical Name or Material Hydrochloric acid

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 4L
    Formula Weight 36.46
    Molecular Formula HCl
    CAS 7647-01-0
    Synonym Muriatic acid solution
    Packaging SAFEMOR Container
    Chemical Name or Material Hydrochloric acid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit