Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, Environmental Grade, 34-37.5%, Alfa Aesar™

single sub-boiling quartz distillation

Manufacturer:  Alfa Aesar 38743AP

Quantity 500mL

 View more versions of this product

Catalog No. AA38743AP


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

7647-01-0
34-37.5%
36.46
Hydrochloric acid, Single sub-boiling quartz distillation
Liquid
HCl
MFCD00011324
Environmental Grade

Specifications

Clear colorless
7647-01-0
Liquid
HCl
MFCD00011324
Pungent
Environmental Grade
500mL
Colorless liquid
34-37.5%
36.46
1.19g/cm3
Hydrochloric acid, Single sub-boiling quartz distillation

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 231-595-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit