Alfa Aesar™ Hydrochloric acid, Environmental Grade Plus, 33-36%

Manufacturer: Alfa Aesar™ 38744AP

Quantity 500mL

 View more versions of this product

Catalog No. AA38744AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-01-0
    Formula Weight 36.46
    MDL Number MFCD00011324
    UN Number UN1789
    Merck Index 14,4780
    Chemical Name or Material Hydrochloric acid
    Grade Environmental Grade Plus

    Specifications

    Physical Form Liquid, Double Sub-Boiling Quartz Distillation
    Formula Weight 36.46
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7647-01-0
    Health Hazard 2 GHS H Statement
    H314-H335
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Boiling Point 81.5° to 110°C
    UN Number UN1789
    Merck Index 14,4780
    Density 1.19g/mL
    Health Hazard 1 Danger
    Odor Pungent
    TSCA TSCA
    MDL Number MFCD00011324
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
    RTECS Number MW4025000
    Quantity 500mL
    Chemical Name or Material Hydrochloric acid
    Assay 33 to 36%
    Grade Environmental Grade Plus

    GHS H Statement
    H314-H335
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit