Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, ICP-OES, for Trace Metal Analysis, Thermo Scientific™

Click to view available options
Quantity:
500 ml
2.5 l
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883

Suitable for digestion of samples prior to trace elemental analysis via ICP-OES. Tested for more than 20 metals at ppb levels and also meets ACS testing specifications.

  • Suitable for trace metal analysis by ICP-OES
  • Tested for more than 20 metal impurities at ppb levels
  • Also meets general ACS testing specifications
  • Ideal for analysis of environmental samples
Quantity 500 ml
Color ≤ 5 (APHA)
Aluminum (Al) ≤ 10 ppb
Ammonium ≤ 3 ppm (ACS)
Antimony (Sb) ≤ 1 ppb
Arsenic (As) ≤ 10 ppb
Barium (Ba) ≤ 10 ppb
Boron (B) ≤ 10 ppb
Cadmium (Cd) ≤ 10 ppb
Calcium (Ca) ≤ 10 ppb
Chromium (Cr) ≤ 10 ppb
Chlorine (Cl) ≤ 500 ppb (ACS)
SMILES [H+].[Cl-]
IUPAC Name hydrogen chloride
Copper (Cu) ≤ 10 ppb
Formula Weight 36.46
Physical Form Liquid
Percent Purity 36.5 to 38.0%
Grade ICP-OES
Heavy Metals (as Pb) ≤ 1 ppm
Iron (Fe) ≤ 10 ppb
Lead (Pb) ≤ 10 ppb
Lithium (Li) ≤ 10 ppb
Manganese (Mn) ≤ 10 ppb
Magnesium (Mg) ≤ 10 ppb
Mercury (Hg) ≤ 1 ppb
Nickel (Ni) ≤ 10 ppb
Packaging PVC-Coated Glass Bottle
Phosphate ≤ 20 ppb (ACS)
Potassium (K) ≤ 10 ppb
Residue after Ignition ≤ 5 ppm (ACS)
Sodium (Na) ≤ 10 ppb
Sulfate ≤ 100 ppb (ACS)
Tin (Sn) ≤ 10 ppb
Titanium (Ti) ≤ 10 ppb
Vanadium (V) ≤ 10 ppb
Zinc (Zn) ≤ 10 ppb
Show More Show Less

RMF - Research Use or Further Mfg