Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, Multi-Compendial, N.F., J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
Glass Bottle, Single ship
pvc coated glass bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
Quantity 2.5L
SMILES Cl
IUPAC Name chlorane
Formula Weight 36.46
Packaging Glass Bottle
Specific Gravity 1L = 1.18kg
Chemical Name or Material Hydrochloric Acid
Grade Multi-Compendial/NF