Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, pure, fuming, 37% solution in water, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
25L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
7647-01-0,7732-18-5
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0,7732-18-5
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
Color Undesignated
Viscosity 1.9 mPa.s (15°C)
Quantity 2.5L
Linear Formula HCl
Fieser 04,450; 05,533; 06,283
Solubility Information Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
Density 1.1800g/mL
Formula Weight 36.46
Physical Form Solution
Percent Purity 35.0 to 39.0%
Grade Pure
Free Chlorine 10ppm max. (Cl)
Iron (Fe) 5ppm max.
Lead (Pb) 2ppm max.
Packaging Glass Bottle
Solubility Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
Specific Gravity 1.18
Sulfate 100ppm max.
Melting Point -35.0°C
Boiling Point 57.0°C
Chemical Name or Material Hydrochloric acid
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger