Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, Reagent, ACS, 36.5-38%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
5 gal.
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Bottle
Poly Drum
Poly-Coated Glass Bottle
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
[H+].[Cl-]
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
[H+].[Cl-]

H1035, 7647-01-0, HCl

Spectrum™ Hydrochloric Acid, Reagent, ACS , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids, it is the least likely to be subject to an oxidation-reduction reaction. The fact that it is available as a pure reagent make it an excellent acidifying reagent. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrochloric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Quantity 200 L
Ammonium 3 ppm
CAS Max % 1
Arsenic (As) 0.01 ppm
Bromide 5e-005
UN Number UN1789
Percent Purity 36.5 to 38%
Heavy Metals (as Pb) 1 ppm
Packaging Drum
Residue after Ignition 5 ppm
Sulfate 1 ppm
Show More Show Less