Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hydrochlorothiazide, 98%

Manufacturer:  Alfa Aesar™ B2209306

Quantity 5g

 View more versions of this product

Catalog No. AAB2209306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

58-93-5
297.74
625101
6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide
C7H8ClN3O4S2
MFCD00051765
14,4781
Hydrochlorothiazide

Specifications

266° to 272°C
58-93-5
C7H8ClN3O4S2
MFCD00051765
14,4781
Hydrochlorothiazide
5g
Odorless
297.74
625101
6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide
98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : DK9100000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit