Alfa Aesar™ Hydrochlorothiazide, 98%

Manufacturer: Alfa Aesar™ B2209322

Quantity 100g

 View more versions of this product

Catalog No. AAB2209322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 58-93-5
    Molecular Formula C7H8ClN3O4S2
    Formula Weight 297.74
    MDL Number MFCD00051765
    Beilstein 625101
    Merck Index 14,4781
    Synonym 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide
    Chemical Name or Material Hydrochlorothiazide

    Specifications

    Formula Weight 297.74
    TSCA TSCA
    Odor Odorless
    Molecular Formula C7H8ClN3O4S2
    MDL Number MFCD00051765
    Merck Index 14,4781
    RTECS Number DK9100000
    Melting Point 266° to 272°C
    Quantity 100g
    CAS 58-93-5
    Beilstein 625101
    Synonym 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide
    Chemical Name or Material Hydrochlorothiazide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit