Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid, 48-50% (Technical), Fisher Chemical

Click to view available options
Quantity:
1 lb.
10 lb.
Packaging:
Poly Bottle
Poly Bottle/HF
7664-39-3,7732-18-5
FH
20.006
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484
14917
CHEBI:29228
fluorane
F
7664-39-3,7732-18-5
FH
20.006
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid,hydrogen fluoride,fluorhydric acid,hydrofluoride,rubigine,fluorwasserstoff,hydrogen-fluoride,antisal 2b,fluorowodor polish,caswell no. 484
14917
CHEBI:29228
fluorane
F
pH 2.0
Quantity 1 lb.
Color Colorless
Assay Percent Range 48 to 50 w/w %
UN Number 1790
Formula Weight 20.01
Physical Form Liquid
Percent Purity 48 to 50% (w/w)
Grade Technical
Identification Pass Test
Packaging Poly Bottle/HF
Melting Point -35°C
Boiling Point 105°C
Chemical Name or Material Hydrofluoric Acid, 48-50%
Show More Show Less

DANGER!

Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:1

DOTInformation : DOT Class 8, : Corrosive