Hydrofluoric acid, ACS, 48-51%, Alfa Aesar™

Manufacturer: Alfa Aesar 3325809

Quantity 10g

 View more versions of this product

Catalog No. AA3325809

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7664-39-3
    Physical Form Liquid
    Assay Percent Range 48-51%
    Molecular Formula HF
    Formula Weight 20.01
    MDL Number MFCD00011346
    Chemical Name or Material Hydrofluoric acid, Special handling precautions required
    Grade ACS Grade

    Specifications

    Formula Weight 20.01
    Quantity 10g
    Color Colorless
    EINECS Number 231-634-8
    Physical Form Liquid
    Assay Percent Range 48-51%
    Molecular Formula HF
    MDL Number MFCD00011346
    CAS 7664-39-3
    Health Hazard 1 H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.
    Appearance Colorless liquid
    Odor Acrid
    Chemical Name or Material Hydrofluoric acid, Special handling precautions required
    Grade ACS Grade

    H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit