Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hydrofluoric acid, Environmental Grade Plus, 47-51%

Manufacturer:  Alfa Aesar™ 38746AU

Quantity 1000mL

 View more versions of this product

Catalog No. AA38746AU


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7664-39-3
MFCD00011346
14,4790
Environmental Grade Plus
20.01
UN1790
Hydrofluoric acid

Specifications

Colorless
7664-39-3
20.01
UN1790
1.16g/mL
112°C
47 to 51%
1000mL
Liquid, Double Sub-Boiling Quartz Distillation
MFCD00011346
14,4790
Acrid
Hydrofluoric acid
Environmental Grade Plus

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID

RTECSNumber : MW7875000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit