Alfa Aesar™ Hydrofluoric acid, Environmental Grade Plus, 47-51%

Manufacturer: Alfa Aesar™ 38746AK

Quantity 250mL

 View more versions of this product

Catalog No. AA38746AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7664-39-3
    Formula Weight 20.01
    MDL Number MFCD00011346
    UN Number UN1790
    Merck Index 14,4790
    Chemical Name or Material Hydrofluoric acid
    Grade Environmental Grade Plus

    Specifications

    Physical Form Liquid, Double Sub-Boiling Quartz Distillation
    Formula Weight 20.01
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 112°C
    MDL Number MFCD00011346
    CAS 7664-39-3
    Density 1.16g/mL
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Odor Acrid
    RTECS Number MW7875000
    Quantity 250mL
    TSCA TSCA
    UN Number UN1790
    Merck Index 14,4790
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID
    Color Colorless
    Chemical Name or Material Hydrofluoric acid
    Assay 47 to 51%
    Grade Environmental Grade Plus

    GHS H Statement
    H300-H310-H330-H314
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit