Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric acid, J.T.Baker™

Click to view available options
Quantity:
2.5L
4L

Chemical Identifiers

7664-39-3
FH
20.006
KRHYYFGTRYWZRS-UHFFFAOYSA-N
Fluoric acid; Fluorhydric acid
14917
CHEBI:29228

High quality chemicals for laboratory and specialized industrial use

    For Microelectronic Use
Color Colorless
Quantity 2.5L
CAS 7664-39-3
Molecular Formula FH
Synonym Fluoric acid; Fluorhydric acid
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Density 1.15g/mL
SMILES F
IUPAC Name fluorane
Molecular Weight (g/mol) 20.006
PubChem CID 14917
ChEBI CHEBI:29228
Formula Weight 36.46
Physical Form Liquid
Specific Gravity 1L = 1.18kg
Melting Point -83.6°C
Boiling Point 19.5°C
Chemical Name or Material Hydrofluoric acid
Assay 48.0 to 51.0%
Grade Baker Analyzed™ ACS Reagent
Show More Show Less