Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrofluoric acid, J.T.Baker™
Click to view available options
Quantity:
2.5L
4L
Description
- For Microelectronic Use
Specifications
Color | Colorless |
Quantity | 4L |
Density | 1.15g/mL |
Formula Weight | 36.46 |
Physical Form | Liquid |
Percent Purity | 48.0 to 51.0% |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Bagged Poly Bottle |
Specific Gravity | 1L = 1.18kg |
Melting Point | -83.6°C |
Show More |