Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging HDPE Bottle
Quantity 4L

 View more versions of this product

Catalog No. A513-4


 

Due to product restrictions, please to purchase or view availability for this product.

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

7664-39-3
20.01
Hydrofluoric Acid
HF
MFCD00011346
TraceMetal

Specifications

Colorless
2.0
Liquid
20.01
1790
HDPE Bottle
−35°C
Hydrofluoric Acid
4L
7664-39-3
HF
MFCD00011346
15, 4827
Certificate of Lot Analysis included with each shipment.
105°C
TraceMetal

Safety and Handling

SDS & Safety Documents

DOTInformation : DOT Class 8, : Corrosive

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit