Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4228983
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
10035-10-6, 64-19-7
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br
10035-10-6, 64-19-7
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Fuming Liquid
Quantity 100 mL
Linear Formula HBr
Flash Point 39°C
Merck Index 11,472
IUPAC Name hydrogen bromide
Formula Weight 80.9
Percent Purity 31 to 34% (HBr)
Grade Pure
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.42
Density 1.4200g/mL
Chemical Name or Material Hydrogen bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 233-113-

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit