Alfa Aesar™ Hydrogen chloride, 1M in acetic acid

Manufacturer: Alfa Aesar™ H27347AU

Quantity 1000mL

 View more versions of this product

Catalog No. AAH27347AU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-01-0
    Molecular Formula HCl
    Formula Weight 36.46
    MDL Number MFCD00011324
    UN Number UN2920
    Chemical Name or Material Hydrogen chloride

    Specifications

    Formula Weight 36.46
    TSCA TSCA
    Quantity 1000mL
    Molecular Formula HCl
    Concentration 1M in acetic acid
    MDL Number MFCD00011324
    UN Number UN2920
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Acrid
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
    Flash Point 40°C (104°F)
    CAS 7647-01-0
    Health Hazard 2 GHS H Statement
    H314-H318-H226-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Chemical Name or Material Hydrogen chloride

    GHS H Statement
    H314-H318-H226-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit