Alfa Aesar™ Hydrogen chloride, 2M in diethyl ether

Manufacturer: Alfa Aesar™ H26914AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH26914AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-01-0
    Molecular Formula HCl
    Formula Weight 36.46
    MDL Number MFCD00011324
    UN Number UN2924
    Chemical Name or Material Hydrogen chloride

    Specifications

    Formula Weight 36.46
    Quantity 100mL
    Concentration 2M in diethyl ether
    UN Number UN2924
    TSCA TSCA
    Flash Point −40°C (−40°F)
    Molecular Formula HCl
    MDL Number MFCD00011324
    CAS 7647-01-0
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H331-H314-H302-H336
    Highly flammable liquid and vapor.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material Hydrogen chloride

    GHS H Statement
    H225-H331-H314-H302-H336
    Highly flammable liquid and vapor.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.

    2° to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit