Alfa Aesar™ Hydrogen chloride, nominally 2.5M in ethanol

Manufacturer: Alfa Aesar™ 44605JT

Quantity 0.5 mole

 View more versions of this product

Catalog No. AA44605JT

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-01-0
    Formula Weight 36.46
    MDL Number MFCD00011324
    UN Number UN2924
    Chemical Name or Material Hydrogen chloride

    Specifications

    Formula Weight 36.46
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H331-H314
    Highly flammable liquid and vapor.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Quantity 0.5 mole
    Physical Form Liquid
    MDL Number MFCD00011324
    CAS 7647-01-0
    UN Number UN2924
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    TSCA TSCA
    Concentration Nominally 2.5M in ethanol
    Chemical Name or Material Hydrogen chloride

    GHS H Statement
    H225-H331-H314
    Highly flammable liquid and vapor.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit