Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, pure, 2N solution in diethyl ether, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
7647-01-0,60-29-7
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0,60-29-7
ClH
36.458
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
Color Colorless
Quantity 800mL
Linear Formula HCl
Fieser 04,450; 05,533; 06,283
Merck Index 10, 4701
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvents
Concentration 1.90 to 2.20N
Density 0.7400g/mL
Formula Weight 36.45
Physical Form Liquid
Grade Pure
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in most organic solvents
Specific Gravity 0.74
Flash Point −40°C
Chemical Name or Material Hydrogen chloride, 2N solution in diethyl ether
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger