Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen chloride, pure, 5 to 6N solution in 2-propanol, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Plastic Bottle
7647-01-0,67-63-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0,67-63-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
Linear Formula HCl
Fieser 04,450; 05,533; 06,283
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
Formula Weight 36.45
Physical Form Liquid
Grade Pure
Concentration 5N min.
Density 0.9090g/mL
Packaging Plastic Bottle
Solubility Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
Specific Gravity 0.909
Water 0.5% max.
Color Colorless to Yellow
Quantity 2.5L
Flash Point 11°C
Chemical Name or Material Hydrogen chloride
Show More Show Less