Learn More
Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180070500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Hydrogen fluoride-pyridine | |
32001-55-1 | |
1.1000g/mL | |
Plastic Bottle in Tin Can | |
50 g | |
16,286; 16,050; 05,538; 06,473; 09,399; 12,419 | |
pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine | |
C1=CC=NC=C1.F | |
99.11 | |
99.11 | |
Liquid |
65-70% | |
Yellow to Brown | |
65-70% | |
C5H6FN | |
MFCD00012436 | |
1.1 | |
GRJJQCWNZGRKAU-UHFFFAOYSA-N | |
pyridine;hydrofluoride | |
64774 | |
65-70% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 250-889-6
TSCA : TSCA
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only