Learn More
Parker Hannifan HYDROGEN GENERATOR DFS Item
Manufacturer:  Parker Hannifan H2PD150
MODEL H2PD-150Parker Fuel and Carrier Gas Hydrogen Generators produce up to 300 cc/min of ultra-high purity (UHP) hydrogen. Applications for hydrogen include use as a fuel gas for Flame Ionization Detectors, as a reaction gas for Hall Detectors, and as a carrier gas to ensure absolute repeatability of retention times.Hydrogen is also used in high sensitivity Trace Hydrocarbon Analyzers and air pollution monitors to ensure the lowest possible background noise. Other applications include hydrogenation reactions and analysis of gas emissionsCategory: Hydrogen Generators for Fuel and Carrier Gas Tags: carrier gas, fid, flame ionization detectors, hall detectors, trace hydrocarbon analyzers, uhp hydrogen
Catalog No. NC1477271