Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen hexachloroiridate(IV) hexahydrate, approx. 40% Iridium, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
16941-92-7
H2Cl6Ir·6H2O
515.02
MFCD00149907
Chloroiridic acid
16941-92-7
H2Cl6Ir·6H2O
515.02
MFCD00149907
Chloroiridic acid
Quantity 5g
Packaging Glass Bottle
Color Black
Linear Formula H2IrCl6·6H2O
Fieser 01,131; 02,67; 03,47; 04,93; 05,119; 05,287; 06,45; 17,269; 05,203; 06,108; 07,59; 08,434; 10,382; 14,294; 17,326
Solubility Information Solubility in water: soluble
Formula Weight 515.02
Physical Form Crystals
Percent Purity approx. 40% Iridium
Chemical Name or Material Hydrogen hexachloroiridate(IV) hexahydrate, Approx. 40% Iridium

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 241-012-8