Alfa Aesar™ Hydrogen hexafluoroantimonate(V), »65% aq. soln.

Manufacturer: Alfa Aesar™ 3348818

Quantity 50g

 View more versions of this product

Catalog No. AA3348818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16950-06-4
    Linear Formula HF·SbF5
    Formula Weight 236.75
    MDL Number MFCD00011327
    UN Number UN3264
    Synonym Hexafluoroantimonic acid; Fluoroantimonic acid
    Chemical Name or Material Hydrogen hexafluoroantimonate(V)

    Specifications

    Formula Weight 236.75
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Linear Formula HF·SbF5
    MDL Number MFCD00011327
    UN Number UN3264
    Synonym Hexafluoroantimonic acid; Fluoroantimonic acid
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Concentration ≈65% aq. soln.
    Color Colorless to dark orange
    Quantity 50g
    TSCA TSCA
    CAS 16950-06-4
    Health Hazard 2 GHS H Statement
    H314-H302-H332
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Hydrogen hexafluoroantimonate(V)

    GHS H Statement
    H314-H302-H332
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit